missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flotillin 2 Recombinant Rabbit Monoclonal Antibody (23GB5435)
GREENER_CHOICE

Catalog No. MA552854
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552854 100 μL
MA552855 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552854 Supplier Invitrogen™ Supplier No. MA552854
Only null left

Rabbit Recombinant Monoclonal Antibody

Flotillin 2 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.

Specifications

Antigen Flotillin 2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5435
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FLOT2
Gene Accession No. Q14254, Q60634, Q9Z2S9
Gene Alias AI573412; ECS1; ECS-1; epidermal surface antigen; ESA; ESA1; FLOT2; flotillin 2; Flotillin 2 (epidermal surface antigen 1); flotillin-2; M17S1; membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; Reg1; REG-1; Reggie-1; reggie1-1; reggie-2
Gene Symbols FLOT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Flotillin 2 (324-366AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14252, 2319, 83764
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less