missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flotillin 1 Recombinant Rabbit Monoclonal Antibody (23GB845)
GREENER_CHOICE

Catalog No. MA554076
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554076 100 μL
MA554077 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554076 Supplier Invitrogen™ Supplier No. MA554076
Only null left

Rabbit Recombinant Monoclonal Antibody

Flotillin 1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.

Specifications

Antigen Flotillin 1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB845
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FLOT1
Gene Accession No. O75955
Gene Alias DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2
Gene Symbols FLOT1
Host Species Rabbit
Immunogen A synthetic peptide of human Flotillin 1 (400-427AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10211
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less