missing translation for 'onlineSavingsMsg'
Learn More

Florfenicol 98.0+%, TCI America™

Catalog No. F08115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08115G 5 g
F08111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08115G Supplier TCI America Supplier No. F08115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73231-34-2
Molecular Formula C12H14Cl2FNO4S
Molecular Weight (g/mol) 358.205
MDL Number MFCD00864834
InChI Key AYIRNRDRBQJXIF-NXEZZACHSA-N
Synonym florfenicol, nuflor, --florfenicol, nuflor gold, florphenicol, aquafen, florfenicol usan:inn:ban, 2,2-dichloro-n-1r,2s-3-fluoro-1-hydroxy-1-4-methylsulfonylphenyl propan-2-yl acetamide, 2,2-dichloro-n-1-fluoromethyl-2-hydroxy-2-4-methylsulfonyl phenyl ethyl acetamide, d-threo-2,2-dichloro-n-alpha-fluoromethyl-beta-hydroxy-p-methylsulfonyl phenethyl acetamide
PubChem CID 114811
ChEBI CHEBI:87185
IUPAC Name 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Formula Weight 358.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Florfenicol
RTECSNumber DB6034000
TSCA No