missing translation for 'onlineSavingsMsg'
Learn More

Florfenicol, MilliporeSigma™ Supelco™

Catalog No. 111015373
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015373 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015373 Supplier Merck Emd Millipore Supplier No. F1427500MG

Florfenicol is a compound, which belongs to the amphenicol class of antibiotics. It is used to treat bacterial infections in food-producing animals.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 73231-34-2
Physical Form Solid
Linear Formula C12H14 Cl2 FNO4 S
MDL Number MFCD00864834
Synonym Aquafen; Nuflor; SCH-25298; [R-(R*,S*)]-2,2-Dichloro-N-[1-(fluoromethyl)-2-hydroxy-2-[4-(methylsulfonyl)phenyl]ethyl]acetamide
Type Analytical Standard
Industry Type Drug Analysis
Quantity 500 mg
Formula Weight 358.21
Grade Analytical Standard