missing translation for 'onlineSavingsMsg'
Learn More

Hughes Floor-Mounted Combination Shower with Closed ABS Bowl
GREENER_CHOICE

Catalog No. 18666668
Change view
Click to view available options
Color:
Green Powder Coat
Polished Brush
Material (Piping Connection):
Galvanized Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material (Piping Connection)
18666668 Green Powder Coat Galvanized Steel
18666669 Polished Brush Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18666668 Supplier Justrite International Supplier No. 18G45GT

Hughes All-in-One Floor Mounted Combination Safety Shower with top water inlet provides initial on-site treatment to eye/face or full body exposure to hazards. Suitable for use in applications that are not subject to the possibility of freezing or overheating

Features:

  • Specially designed valve that includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure
  • Low operating torque offers reliable, easy on/off operation. Durable, coated stainless steel pull handle with nylon valve connection is easy to grip and activate for immediate water drenching
  • Designed for high flow rates that meet or exceed ANSI standards, the nozzle offers optimal spray distribution for user protection
  • Unlike shower rose designs, this compact nozzle reduces the potential for insect nesting and eliminates high replacement costs associated with cracked or dented plastic spray heads.
  • Made of durable, corrosion-resistant acrylic capped ABS, the eye/face wash with high, ANSI-compliant flow rate features an integral lid connected to the water valve for automatic operation—activate by simply pulling down the lid
  • Easy-to-reach flow regulator valve positioned on the side of the bowl adjusts to suit the severity of the accident.
  • Solid construction for added corrosion resistance.
  • Includes an inspection record tag for recording service and test history
  • Covered bowl ensures eyewash nozzles and bowl remain clean and free from contamination
  • Twin aerated diffusers provide a soft, scrubbing action to the eyes and face
  • Removable, self-draining nozzle constructed from durable nylon
  • High-performance quarter-turn ball valve made of 316L stainless steel endure the harshest environments and site conditions
  • Includes a universal eye wash and safety shower sign for enhanced visibility
  • Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements

Compliance

ANSI Z358.1-2014 and EN15154 Compliant

Warranty

1 Year

Specifications

Activation Shower: Hand Lever, Eye and Face Wash: Pull Lid Forward
Bowl Size (Metric) 241 mm
Certifications/Compliance ANSI Z358.1-2014 and EN15154 Compliant
Color Green Powder Coat
Depth (English) 16 in.
Depth (Metric) 406 mm
Flow Rate 3 gpm
For Use With (Application) Emergency Shower and Eye Wash with Water
Height (English) 16 in.
Height (Metric) 406 mm
Includes 1 Unit
Inlet Connection (Metric) 1.25 in. (F) NPT
Material Nylon 6, Chrome-plated Brass, Stainless Steel Mesh
Material (Piping Connection) Galvanized Steel
Mounting Floor
Pressure 29 to 87 psi
Product Type Safety Shower and Emergency Eye Wash
Width (English) 48 in.
Width (Metric) 1219 mm
Show More Show Less