Learn More
Thermo Scientific™ FlipStrip™ Microtube Racks with Lid
GREENER_CHOICE
Hold 0.2mL strips and 1.5mL tubes on one side and and 0.5mL tubes on the other side of this microtube rack with lid, which is made from 100% recycled plastic.
$119.64
Specifications
Material Recycled Plastic
Stackable Yes
No. per Case 12 Racks
Product Type Flipstrip Rack
View More Specs
Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
2140237
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
8688
Fluorescent Green
Each for $119.64
Case of 12 Each for $1,435.68
Only null left
Description
Description
Hold 0.2mL strips and 1.5mL tubes on one side of the Thermo Scientific™ FlipStrip Microtube Rack with Lid, and 0.5mL tubes on the other. The rack is made from 100% recycled plastic.
  • Stable, low-profile design with an alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
  • Several colors available
Specifications
Specifications
Recycled Plastic
Yes
12 Racks
Flipstrip Rack