Learn More
Fisherbrand™ Flexelene Tubing
General use bioprocess and medical grade tubing. UV stable and suitable for use with deionized water. USP Class VI tubing, withstands EtO sterilization and gamma sterilization to 255 kGy. Shore A 86.
 View more versions of this product
Catalog No. 01000779
Only null left
Description
Description
  • PVC-Free - No DEHP or BPA additives
  • Low extractables / leachables
  • ISO 10993-5 – non-cytotoxic
  • ISO 10993-4 – non-hemolytic
  • Non-Animal Derived
  • RoHS Compliant
  • Lot Traceable
Warranty
Replacement if not satisfied. 12 Month return Policy.
Specifications
Chemical Identifiers
Complaince Certificates Available
100 ft. Roll - Bag
Out of direct sunlight, >20°F <100°F
Tubing
Multi-purpose Medical, Pharmaceutical uses
Flexelene™ Tubing
Transparent
Specifications
Complaince Certificates Available
Polyolefin Elastomer
Multi-purpose Medical, Pharmaceutical uses
Flexelene™ Tubing
Transparent
1/4 in.
3/8 in.
100 ft. Roll - Bag
Out of direct sunlight, >20°F <100°F
Tubing
Product Suggestions
Product Suggestions