Learn More
Fisherbrand™ Flexelene 135C
A bioprocess and medical grade tubing that meets USP Class VI requirements as well as cytotoxicity, ISO 10993-5 and hemolysis, ISO 10993-4 requirements. The heat-sealable and thermally weldable TPE polymer is autoclavable to 121°C and has the durability required for use in peristaltic pumps.
 View more versions of this product
Catalog No. 01000748
Only null left
Description
Description
  • PVC-Free - No DEHP or BPA additives
  • Ultra-low extractables / leachables
  • ISO 10993-5 – non-cytotoxic
  • ISO 10993-4 – non-hemolytic
  • Low gas and oxygen permeability
  • Low protein binding
  • Non-pyrogenic material
  • Thermally weldable
  • Non-Animal Derived
  • RoHS Compliant
  • Lot Traceable
Warranty
Replacement if not satisfied. 12 Month return Policy.
Specifications
Chemical Identifiers
Complaince Certificates Available
TPE
50 ft. Roll - Bag
Out of direct sunlight, >20°F <100°F
Tubing
Class 7 Cleanroom Manufactured
Medical, Pharmaceutical Uses; Peristaltic Pump Tubing
Flexelene™ 135C Tubing
White Translucent
Specifications
Complaince Certificates Available
3/16 in.
5/16 in.
50 ft. Roll - Bag
Out of direct sunlight, >20°F <100°F
Tubing
Class 7 Cleanroom Manufactured
TPE
Medical, Pharmaceutical Uses; Peristaltic Pump Tubing
Flexelene™ 135C Tubing
White Translucent
Product Suggestions
Product Suggestions