missing translation for 'onlineSavingsMsg'
Learn More

Fleroxacin 98.0+%, TCI America™

Catalog No. F06465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06465G 5 g
F064625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06465G Supplier TCI America Supplier No. F06465G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79660-72-3
Molecular Formula C17H18F3N3O3
Molecular Weight (g/mol) 369.34
MDL Number MFCD00864880
InChI Key XBJBPGROQZJDOJ-UHFFFAOYSA-N
Synonym 6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid, Megalone
PubChem CID 3357
ChEBI CHEBI:31810
IUPAC Name 6,8-difluoro-1-(2-fluoroethyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
SMILES CN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(CCF)C2=C1F)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 369.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Fleroxacin
RTECSNumber VB1999050
TSCA No