CAS RN 79660-72-3
IUPAC Name:
6,8-difluoro-1-(2-fluoroethyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms:
fleroxacin quinodis FLRX fleroxicin 氟罗沙星 megalone grünenthal brand of fleroxacin fleroxacin (substance) fleroxacino fleroxacine
Molecular Weight (g/mol):
369.34
Molecular Formula:
C17H18F3N3O3
InChi Key:
XBJBPGROQZJDOJ-UHFFFAOYSA-N
SMILES:
CN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(CCF)C2=C1F)C(O)=O
1
–
1
of
1
results
1
Fleroxacin 98.0+%, TCI America™
CAS: 79660-72-3 Molecular Formula: C17H18F3N3O3 Molecular Weight (g/mol): 369.34 MDL Number: MFCD00864880 InChI Key: XBJBPGROQZJDOJ-UHFFFAOYSA-N Synonym: 6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid, Megalone PubChem CID: 3357 ChEBI: CHEBI:31810 IUPAC Name: 6,8-difluoro-1-(2-fluoroethyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid SMILES: CN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(CCF)C2=C1F)C(O)=O