missing translation for 'onlineSavingsMsg'
Learn More

Flavoxate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7129851
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07175G 5 g
F071725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07175G Supplier TCI America Supplier No. F07175G
Only null left

Chemical Identifiers

CAS 3717-88-2
Molecular Formula C24H26ClNO4
Molecular Weight (g/mol) 427.925
MDL Number MFCD00072099
InChI Key XOEVKNFZUQEERE-UHFFFAOYSA-N
PubChem CID 441345
ChEBI CHEBI:5089
IUPAC Name 2-piperidin-1-ylethyl 3-methyl-4-oxo-2-phenylchromene-8-carboxylate;hydrochloride
SMILES CC1=C(OC2=C(C1=O)C=CC=C2C(=O)OCCN3CCCCC3)C4=CC=CC=C4.Cl

Specifications

Melting Point 234°C
Color White-Yellow
Quantity 5 g
Formula Weight 427.93
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Flavoxate Hydrochloride
RTECSNumber DJ2450000
TSCA No