accessibility menu, dialog, popup

UserName

HClH3COOOON

CAS RN 3717-88-2

IUPAC Name: hydrogen 2-(piperidin-1-yl)ethyl 3-methyl-4-oxo-2-phenyl-4H-chromene-8-carboxylate chloride
Synonyms: hydrogen flavoxate chloride
Molecular Weight (g/mol): 427.93
Molecular Formula: C24H26ClNO4
InChi Key: XOEVKNFZUQEERE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=C(OC2=C(C=CC=C2C1=O)C(=O)OCCN1CCCCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)