missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flavivirus Envelope Recombinant Mouse Monoclonal Antibody (D1-4G2-4-15 (4G2))
GREENER_CHOICE

Catalog No. MA551803
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551803 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551803 Supplier Invitrogen™ Supplier No. MA551803
Only null left

Mouse Recombinant Monoclonal Antibody

Recognises flavivirus group specific antigens (Dengue virus, West Nile Virus, Japanese Encephalitis, Yellow Fever Virus, Zika virus etc). It binds to the fusion loop at the extremity of domain II of protein E.

The Flaviviridae are a family of viruses that are primarily spread through arthropod vectors (mainly ticks and mosquitoes). Flaviviridae have monopartite, linear, single-stranded RNA genomes of positive polarity, 9.6- to 12.3-kilobase in length. Virus particles are enveloped and spherical, about 40-60 nm in diameter.

Specifications

Antigen Flavivirus Envelope
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Neutralization, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone D1-4G2-4-15 (4G2)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Host Species Mouse
Immunogen Dengue Virus type 2 antigens.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less