missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Flat Bottom Glass Crystallizing Dishes with Spout

Flat bottom glass crystallizing dishes with spouts for easy, mess free pouring. Ideal for storage and crystallization. Reusable and autoclavable.

Supplier:  Eisco™ CH0066I

 View more versions of this product

Catalog No. S27495


Only null left

Description

Description

  • Made from high quality borosilicate 3.3 glass
  • Spout for easy, mess free pour
  • Autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Chemical Identifiers

Autoclavable
Clear
Non-sterile
Crystallizing Dish
Flat
Borosilicate Glass
Non-treated

Specifications

16.9 oz.
Flat
Clear
Cylindrical
Non-treated
500 mL
1 Ea.
63.5 mm
4.5 in.
Autoclavable
500 mL
Borosilicate Glass
Non-sterile
Crystallizing Dish
Crystallization
1/Pk.
2.5 in.
115 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions