Learn More
Eisco™ Flat Bottom Glass Crystalizing Dishes
Eisco™ Flat Bottom Glass Crystalizing Dishes have no spout and are ideal for storage and crystallization.
Supplier:  Eisco™ CH0067D
 View more versions of this product
Catalog No. S88370
Only null left
Description
Description
  • Made from high quality Borosilicate 3.3 Glass
  • Ideal for storage and crystallization
  • No spout
  • Reusable
  • Autoclavable
Specifications
Chemical Identifiers
Autoclavable
Clear
Non-sterile
Crystallizing Dish
Flat
Round
Non-treated
Heating baths, cooling baths, crystallization, evaporation
Specifications
3.4 oz.
Flat
Clear
Round
Non-treated
100 mL
70 mm
1.5 in.
71 mm
Autoclavable
100 mL
Borosilicate Glass
Non-sterile
Crystallizing Dish
Heating baths, cooling baths, crystallization, evaporation
40 mm
2.8 in.
Product Suggestions
Product Suggestions