Learn More
Eisco™ Flat Bottom Glass Crystalizing Dishes
Eisco™ Flat Bottom Glass Crystalizing Dishes have no spout and are ideal for storage and crystallization.
Supplier:  Eisco™ CH0067B
 View more versions of this product
Catalog No. S88368
Only null left
Description
Description
  • Made from high quality Borosilicate 3.3 Glass
  • Ideal for storage and crystallization
  • No spout
  • Reusable
  • Autoclavable
Specifications
Chemical Identifiers
Autoclavable
Clear
Non-sterile
Crystallizing Dish
Flat
Round
Non-treated
Heating baths, cooling baths, crystallization, evaporation
Specifications
1.4 oz.
Flat
Clear
Round
Non-treated
40 mL
2 in.
5 cm
1.1 in.
49 mm
Autoclavable
40 mL
Borosilicate Glass
Non-sterile
Crystallizing Dish
Heating baths, cooling baths, crystallization, evaporation
50 mm
30 mm
1.9 in.
Product Suggestions
Product Suggestions