missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Clips and Fasteners for Precision™ Water Baths
Use these flask clips in water baths.
Supplier: Thermo Scientific™ 3166198
Description
Clips and fasteners for use with Thermo Scientific™ Precision™ water baths.
Features:
Flask Clips
- Secure various size flasks to bath platform
- Stainless steel
- Each requires one fastener (Cat. No. 3166189 - order separately)
Test Tube Clip
- Stainless steel
- Hold 13 mm to 25 mm tubes
- Each requires one fastener (Cat. No. 3166189 - order separately)
Warranty: varies by country, please contact us for details
Chemical Identifiers
| For use in Precision Reciprocal, Dubnoff, and Shallow-Form Shaking Water Baths. | |
| Reciprocating |
Specifications
| 50 mL | |
| Reciprocating | |
| 15 x 50 mL (Model 25), 36 x 50 mL (Model 50) Flasks |
| For use in Precision Reciprocal, Dubnoff, and Shallow-Form Shaking Water Baths. | |
| Flask Clip |