missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clips and Fasteners for Precision™ Water Baths

Use these flask clips in water baths.

Supplier:  Thermo Scientific™ 3166221

 View more versions of this product

Catalog No. 1545515D


Only null left

Description

Description

Clips and fasteners for use with Thermo Scientific™ Precision™ water baths.

Features:

Flask Clips

  • Secure various size flasks to bath platform
  • Stainless steel
  • Each requires one fastener (Cat. No. 3166189 - order separately)

Test Tube Clip

  • Stainless steel
  • Hold 13 mm to 25 mm tubes
  • Each requires one fastener (Cat. No. 3166189 - order separately)

Warranty: varies by country, please contact us for details

Specifications

Chemical Identifiers

For use in Precision Reciprocal, Dubnoff, and Shallow-Form Shaking Water Baths.
Reciprocating

Specifications

125 mL
Reciprocating
9 x 125 mL (Model 25), 24 x 125 mL (Model 50) Flasks
For use in Precision Reciprocal, Dubnoff, and Shallow-Form Shaking Water Baths.
Flask Clip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions