missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Amino
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
$222.16 - $1112.30
Specifications
| Material | Polypropylene |
|---|---|
| Type | Amino |
| Resolution | 40 to 60 μm |
| Column Type | Amino |
| For Use With (Application) | Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates |
| Catalog Number | Mfr. No. | Size (Metric) Tube Rack | Includes | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size (Metric) Tube Rack | Includes | Price | Quantity | ||||
|
05410038
|
BUCHI
140000064 |
120 g | 2 Pc. |
Pack of 2 Each for $782.16
|
|
||||
|
05410034
|
BUCHI
140000060 |
12 g | 5 Pc. |
Pack of 5 Each for $359.46
|
|
||||
|
05410035
|
BUCHI
140000061 |
20 g | 2 Pc. |
Pack of 2 Each for $236.04
|
|
||||
|
05410039
|
BUCHI
140000065 |
220 g | 1 Pc. |
Pack for $766.73
|
|
||||
|
05410040
|
BUCHI
140000066 |
330 g | 1 Pc. |
Pack for $1,112.30
|
|
||||
|
05410036
|
BUCHI
140000062 |
40 g | 2 Pc. |
Pack of 2 Each for $296.20
|
|
||||
|
05410033
|
BUCHI
140000059 |
4 g | 5 Pc. |
Pack of 5 Each for $222.16
|
|
||||
|
05410037
|
BUCHI
140000063 |
80 g | 2 Pc. |
Pack of 2 Each for $536.88
|
|
||||
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
| Polypropylene | |
| 40 to 60 μm | |
| Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates |
| Amino | |
| Amino | |
| Any Flash System |