Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Amino
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
Supplier: BUCHI 140000065
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
| Polypropylene | |
| 1 Pc. | |
| 220 g | |
| Better Peak Shapes with Polar and Basic Compounds, Ideal for Carbohydrates |
| Amino | |
| 40 to 60 μm | |
| Amino | |
| Any Flash System |