Learn More
Fisherbrand™ Flange Plug Caps, 13 mm
Catalog No. 02681509
Change view
Click to view available options
Color:
Black
Blue
Green
Lavender
Natural
Orange
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02681509 Blue
02681510 Natural
02681511 Green
02681512 Black
02681513 Lavender
02681514 Orange
02681515 Red
02681516 White
02681517 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02681509 Supplier Fisherbrand™ Supplier No. 118240BPLFH
Only null left
These polyethylene (PE) flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes and round cuvettes.
  • Two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps.
Specifications
Closure Size 13 mm
Autoclavable Not Autoclavable
Material Polyethylene
Color Blue
Quantity 1000/Cs.