Learn More
Fisherbrand™ Flange Plug Caps, 12 mm
Catalog No. 02681508
Change view
Click to view available options
Color:
Black
Blue
Green
Lavender
Natural
Orange
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02681508 Yellow
02681500 Blue
02681501 Natural
02681502 Green
02681503 Black
02681504 Lavender
02681505 Orange
02681506 Red
02681507 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02681508 Supplier Fisherbrand™ Supplier No. 118127YPLFH
These polyethylene (PE) flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes and round cuvettes.
  • Two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps.
Specifications
Closure Size 12 mm
Autoclavable Not Autoclavable
Material Polyethylene
Color Yellow
Quantity 1000/Cs.
Sterility Non-sterile
Disposable Single-use