missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FKBP8 Recombinant Rabbit Monoclonal Antibody (23GB5890)

Catalog No. MA552941
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552941 20 μL
MA552940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552941 Supplier Invitrogen™ Supplier No. MA552941
Only null left

Rabbit Recombinant Monoclonal Antibody

FKBP8 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.

Specifications

Antigen FKBP8
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5890
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FKBP8
Gene Accession No. Q14318
Gene Alias 38 kDa FK506-binding protein; 38 kDa FKBP; 38kDa; FK506 binding protein 8; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP prolyl isomerase 8; FKBP38; FKBP-38; FKBP8; FKBP-8; FKBPR38; hFKBP38; mFKBP38; peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; rotamase; Sam11
Gene Symbols FKBP8
Host Species Rabbit
Immunogen A synthesized peptide derived from human FKBP38 (1-46AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23770
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less