missing translation for 'onlineSavingsMsg'
Learn More

Edvotek™ Fixed Volume MiniPipets

Catalog No. S90244J
Encompass
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S90244J 100 μL
S90244G 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S90244J Supplier Edvotek™ Supplier No. EPD5883

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easy-to-use, precise operation and ergonomic design

  • Utilize standard disposable micropipet tips
  • Lifetime warranty
  • Cost-effective

Warranty

Lifetime

Specifications

Compatible Tips Standard micropipet tip
Autoclavable Not Autoclavable
Color Gray
Disposable Single-use
For Use With Standard Micropipette Tip
Material Plastic
Model Fix 100 μL
Number of Channels 1
Sterility Non-sterile
Type Fixed Volume Minipipet
Volume (Metric) 100 μL
For Use With (Equipment) Standard micropipet tip
Warranty Lifetime
Show More Show Less