missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FITC Chimeric Recombinant Rat Monoclonal Antibody (4-4-20 (enhanced))

Rat Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551714
Description
This antibody is specific for fluorescein I.
FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs ultraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.Chemical Identifiers
| FITC Chimeric | |
| 1 mg/mL | |
| Functional Assay | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 0.02% ProClin 300 | |
| Fluorescein I-keyhole limpet hemocyanin (FlI-KLH). | |
| Antibody |
| 4-4-20 (enhanced) | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Chemical | |
| Fluorescein | |
| Primary | |
| Protein A |
Specifications
| FITC Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Rat | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2a κ |
| Functional Assay | |
| 4-4-20 (enhanced) | |
| Unconjugated | |
| Fluorescein | |
| Fluorescein I-keyhole limpet hemocyanin (FlI-KLH). | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |