missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FITC Chimeric Recombinant Mouse Monoclonal Antibody (4-4-20 (enhanced))
GREENER_CHOICE

Catalog No. MA551703
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551703 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551703 Supplier Invitrogen™ Supplier No. MA551703
Only null left

Mouse Recombinant Monoclonal Antibody

This antibody is specific for fluorescein I.

FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs ultraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.

Specifications

Antigen FITC Chimeric
Applications Functional Assay
Classification Recombinant Monoclonal
Clone 4-4-20 (enhanced)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias Fluorescein
Host Species Mouse
Immunogen Fluorescein I-keyhole limpet hemocyanin (FlI-KLH).
Purification Method Metal-chelate chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype F(ab) κ
Show More Show Less