Learn More
Fisherbrand™ Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
Description
Zipper-locking feature securely seals bags
- Durable, transparent polyethylene
- Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
- Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
| Not Autoclavable | |
| Zip Seal | |
| LDPE | |
| 100 | |
| 3 x 2 in. (7 x 5cm) | |
| 7 cm | |
| 5 cm |
| 5 oz. | |
| Clear | |
| 1000 | |
| 2 mil | |
| 3 in. | |
| 2 in. |