Learn More
Fisherbrand™ Wash Bottles with Adjustable Sprayer
Handle hot/cold, aqueous/solvent-based solutions
$93.69 - $150.55
Specifications
Color White
Includes Polypropylene adjustable sprayer
Material HDPE
Sterility Non-sterile
Closure Included Yes
View More Specs
Products 2
Catalog Number Capacity (Metric) Price Quantity & Availability  
Catalog Number Capacity (Metric) Price Quantity & Availability  
03405A
View Documents Promotion Details
240 mL
Pack of 3 Each for $93.69
Only null left
03405
View Documents Promotion Details
1000 mL
Each for $150.55
Only null left
Description
Description
Handle hot/cold, aqueous/solvent-based solutions using Fisherbrand™ Wash Bottles with Adjustable Sprayer.
Fully adjustable spray pattern: 'fine mist' for TLC plates; 'needle' for rinsing glassware; 'big stream' for powerful jet needed to wash filters.
  • Bottle is made of high-density polyethylene
  • Sprayer is polypropylene
  • Trigger allows one-handed, controlled delivery with light pressure
  • Engraved markings show how much fluid is left
  • Squat shape takes up little bench space and ensures a low center of gravity
Specifications
Specifications
White
HDPE
Yes
No
Squat
Spray Bottle
Polypropylene
Not Autoclavable
Polypropylene adjustable sprayer
Non-sterile
Reusable
Threaded
10.16 cm
4 in.
Threaded