missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vial Screw Caps

Catalog No. 22045547
Change view
Click to view available options
Color:
Blue
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22045547 Blue
22045548 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22045547 Supplier Fisherbrand™ Supplier No. T501B

Vial Screw Caps are for externally threaded sample tubes and feature long skirts and a super-fast thread design that allows them to be removed or sealed with single turn.

  • Made of polypropylene
  • With lip seal design for leakproof seal
  • Autoclavable
  • For externally threaded tubes
  • Tubes sold separately

Specifications

Color Blue
Material Polypropylene
Septum No
For Use With (Application) External thread sample tubes (T501)
Closure Material Polypropylene
Quantity 1000 Pk.
Closure Size 1.5 to 5 mL tubes
Product Type Screw Cap