missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ VersaClosure™ Tube Closures
GREENER_CHOICE

Catalog No. 0270722
Change view
Click to view available options
Color:
Black
Blue
Clear
Gray
Green
Lavender
Red
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
0270722 Gray
0270717 Clear
0270718 Red
0270719 Lavender
0270720 Green
0270721 Blue
0270723 Yellow
0270724 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 0270722

Universal cap closure

  • Firm, positive seal by sealing either inside the tube or over the tube rim
  • One-size design seals most 16, 13, 12 and 10mm blood collection, glass and plastic test tubes
  • No need to purchase size-specific closures
  • Help with laboratory standardization and managed cost initiatives
  • One-hand technique, seal under various temperature/storage conditions, help preserve sample integrity and minimize evaporation loss

Specifications

Autoclavable Not Autoclavable
Material LDPE
Color Gray
Sterility Non-sterile
For Use With (Equipment) 16, 13, 12 and 10 mm Blood Collection, Glass and Plastic Test Tube