Learn More
Fisherbrand™ VersaClosure™ Tube Closures
GREENER_CHOICE
Universal cap closure
 View more versions of this product
Catalog No. 0270719
Only null left
Description
Description
  • Firm, positive seal by sealing either inside the tube or over the tube rim
  • One-size design seals most 16, 13, 12 and 10mm blood collection, glass and plastic test tubes
  • No need to purchase size-specific closures
  • Help with laboratory standardization and managed cost initiatives
  • One-hand technique, seal under various temperature/storage conditions, help preserve sample integrity and minimize evaporation loss
Specifications
Chemical Identifiers
Not Autoclavable
LDPE
Non-sterile
16, 13, 12 and 10 mm Blood Collection, Glass and Plastic Test Tube
Specifications
Not Autoclavable
Lavender
16, 13, 12 and 10 mm Blood Collection, Glass and Plastic Test Tube
LDPE
Non-sterile
Product Suggestions
Product Suggestions