missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Universal Test Tube Plug Caps
GREENER_CHOICE

Catalog No. 21235720
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21235720 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21235720 Supplier Fisherbrand™ Supplier No. M995135
Only null left

For plastic or glass test tubes.

Innovative flanged-thread design allows cap to descend into the tube until a friction fit is secure, and plug is stationary.

  • Fit tubes with 12 to 16mm diameter
  • Hollow top finish with ribs for easy retrieval

Specifications

Product Type Test Tube Plug Cap
Material LDPE
Color Yellow
No. per Case 1000
Quantity 1000/Cs.
Sterility Non-sterile
For Use With (Equipment) Fits Tubes with Inner Diameters of 12 to 16 mm