Learn More
Fisherbrand™ Three-hole Twistit™ Rubber Stoppers
Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
$158.54 - $175.36
Specifications
Material Natural Rubber
Product Type Stopper (3 holes)
For Use With (Application) Multi-purpose applications
Total Positions 3
View More Specs
Products 11
Catalog Number Color Stopper Number Price Quantity & Availability  
Catalog Number Color Stopper Number Price Quantity & Availability  
14131C
View Documents Promotion Details
Black 5
Pack of 35 Each for $158.54
Case of 10 Pack for $1,585.40
Only null left
14131D
View Documents Promotion Details
Black 6
Pack of 25 Each for $158.54
Case of 10 Pack for $1,585.40
Only null left
14131E
View Documents Promotion Details
Black 7
Pack of 20 Each for $158.54
Case of 10 Pack for $1,585.40
Only null left
14131F
View Documents Promotion Details
Black 8
Pack of 16 Each for $158.54
Case of 10 Pack for $1,585.40
Only null left
14131G
View Documents Promotion Details
Black 9
Pack of 12 Each for $158.54
Case of 10 Pack for $1,585.40
Only null left
14131H
View Documents Promotion Details
Black 10
Pack of 10 Each for $158.54
Case of 10 Pack for $1,585.40
Only null left
14132D
View Documents Promotion Details
White 6
Pack of 25 Each for $175.36
Case of 10 Pack for $1,753.60
Only null left
14132E
View Documents Promotion Details
White 7
Pack of 20 Each for $175.36
Case of 10 Pack for $1,753.60
Only null left
14132H
View Documents Promotion Details
White 10
Pack of 10 Each for $175.36
Case of 10 Pack for $1,753.60
Only null left
14132F
View Documents Promotion Details
White 8
Pack of 16 Each for $175.36
Case of 10 Pack for $1,753.60
Only null left
14132G
View Documents Promotion Details
White 9
Pack of 12 Each for $172.54
Case of 10 Pack for $1,725.40
Only null left
Description
Description
  • Simple twist-off seals provide convenient one, two, or three-hole use
  • Solid stoppers are vacuum-tested at 30 in. of mercury
  • High resistance to deformation—even after repeated autoclaving
  • Each stopper has its size number in raised figures on top
Specifications
Specifications
Natural Rubber
Stopper (3 holes)
Multi-purpose applications
3