missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Three-hole Twistit™ Rubber Stoppers

Catalog No. 14132H
Change view
Click to view available options
Color:
Black
White
Stopper Number:
10
5
6
7
8
9
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Stopper Number
14132H White 10
14131C Black 5
14131D Black 6
14131E Black 7
14131F Black 8
14131G Black 9
14131H Black 10
14132D White 6
14132E White 7
14132F White 8
14132G White 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 14132H

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation

  • Simple twist-off seals provide convenient one, two, or three-hole use
  • Solid stoppers are vacuum-tested at 30 in. of mercury
  • High resistance to deformation—even after repeated autoclaving
  • Each stopper has its size number in raised figures on top

Specifications

Product Type Stopper (3 holes)
Stopper Number 10
Total Positions 3
Size 10
Material Natural Rubber
Color White
For Use With (Application) Multi-purpose applications