missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ Conductivity Calibration Standards Certified Reference Material (CRM), Fisherbrand™

Catalog No. 093281
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
093281 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 093281

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use NIST-traceable reference materials for the highest accuracy available. TraceableTM Conductivity Calibration Standards Certified Reference Material (CRM) offer easy calibration with included instructions and temperature chart.

Traceable Conductivity Calibration Standards Certified Reference Materials (CRM) offer easy calibration with included instructions and temperature chart. For the highest accuracy available, use NIST-traceable reference materials.

Each bottle includes a NIST-traceable calibration report supplied by the manufacturer and information from batch testing. Also includes step-by step calibration instructions and temperature compensation chart. Accuracy at 25°C is (±0.25 microsiemens for 1, 5, 10 microsiemen solution or ±0.25% for other solutions) or the uncertainty shown on the certificate, whichever is greater.

Specifications

Chemical Name or Material Conductivity Standard
Packaging Bottle
Conductivity 10 μS/cm @ 25°C
Technique Calibration of Conductivity Instrument
Color Clear
pH 7 to 9
For Use With (Equipment) Conductivity Meter
Solubility Soluble in water
Solution Type Calibration Solution
Type Conductivity Standard
Quantity 16 oz.
Show More Show Less