Learn More
Fisherbrand™ Top-Opening Five-Slide Plastic Microscope Slide Mailer and Transporter
Use this 50-gauge polypropylene slide container for air conveyors and specimen slide transport
Catalog No. 12-587-17B
Only null left
Explore available promotions
Description
Description
Use this 50-gauge polypropylene slide container for air conveyors and specimen slide transport
  • Holds up to five 3 × 1 in. (75 × 25mm) slides
  • Ribbed partitions keep slides apart
  • Slides not included
Specifications
Specifications
Slide Mailer
3 in. x 1 in.
5 Slides
Product Suggestions
Product Suggestions