Learn More
Fisherbrand™ Surface Treated Tissue Culture Dishes
Catalog No. FB012925
Change view
Click to view available options
Diameter (Metric):
35 mm
60 mm
70 mm
90 mm
100 mm
150 mm
Culture Area:
143 sq. cm
21.2 sq. cm
36.3 cm2
55 sq. cm
60.8 sq. cm
8.5 sq. cm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Culture Area
FB012925 150 mm 143 sq. cm
FB012922 70 mm 36.3 cm2
FB012923 90 mm 55 sq. cm
FB012920 35 mm 8.5 sq. cm
FB012921 60 mm 21.2 sq. cm
FB012924 100 mm 60.8 sq. cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. FB012925
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Containers for cell growth, aids in sample preparation, pretreatment, storage.
  • Flat bottom uniform wall thickness to ensure distortion free bottom
  • Smooth, free surface
  • Less striation to maximize usable area for growth
  • Rim on upper side of lid mates with dish brim for easy, secure stack
  • Lids with chimbs to shape vents
  • Sterilized by gamma irradiation
  • Non-pyrogenic
Warranty
Three years
Specifications
Closure Included Closure Included
Closure Material Polystyrene
Description Cell and tissue culture dishes
Bottom Shape Flat
Culture Area 143 sq. cm
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Standard
Type Cell Culture Dish
For Use With (Application) Tissue culture, storage
Includes Lid
No. per Case 120 Plates
No. per Pack 1 Plate
Dimensions (L x W) 21.9 x 143 mm
Diameter (Metric) 150 mm
Show More Show Less