missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Superfrost™ Slides

Catalog No. 22310398
Change view
Click to view available options
Color:
Clear
Green
Length (Metric):
75 mm
Width (Metric):
25 mm
50 mm
Quantity:
144 Pk., 10 Pk./Cs.
144/Pk., 5 Pk./Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (Metric) Width (Metric) Quantity
22310398 Green 75 mm 25 mm 144 Pk., 10 Pk./Cs.
1255014G Clear 75 mm 25 mm 144 Pk., 10 Pk./Cs.
12550021 Clear 75 mm 50 mm 144/Pk., 5 Pk./Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22310398 Supplier Fisherbrand™ Supplier No. 6941F006
Only null left

Durable and opaque

  • Autoclavable
  • Available in an array of tab colors
  • Offered in standard and large sizes, for positioning larger specimens
  • Durable, opaque tab is inert to common laboratory chemicals and reagents
  • Packaged in paperboard box

Specifications

Length (Metric) 75 mm
Frosted SuperFrost
Width (Metric) 25 mm
Thickness (Metric) 1 mm
Color Green
Length (English) 3 in.
Quantity 144 Pk., 10 Pk./Cs.
Width (English) 1 in.
Dimensions (L x W) 3 x 1 in. (75 x 25 mm)