missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Superfrost™ Slides

Catalog No. 22034980
Change view
Click to view available options
25 x 75 x 1mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22034980 25 x 75 x 1mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22034980 Supplier Fisherbrand™ Supplier No. 4941I2997600621

  • Autoclavable
  • Available in an array of tab colors
  • Offered in standard and large sizes, for positioning larger specimens
  • Durable, opaque tab is inert to common laboratory chemicals and reagents
  • Packaged in paperboard box

Specifications

Length (Metric) 75 mm
Frosted SuperFrost
Width (Metric) 25 mm
Thickness (Metric) 1 mm
Color Clear
Length (English) 3 in.
Quantity 144 Pk., 10 Pk./Cs.
Width (English) 1 in.
Dimensions (L x W) 3 x 1 in. (75 x 25 mm)