missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Superfrost™ Disposable Microscope Slides
Superfrost coating impervious to most common laboratory chemicals and reagents
Description
Superfrost coating impervious to most common laboratory chemicals and reagents
- Brilliant white, opaque coating takes and retains most laboratory marking methods
- Each side has a written key for easy orientation; the word “SPECIMEN” instantly tells you which side is up
- Slides can be imprinted with your laboratory or hospital's name (call your Fisher HealthCare™ Sales Representative for details)
Order Info
Come in handy one-at-a-time dispenser boxes of 72 slides.
Specifications
| Disposable Microscope Slide | |
| 1.2 mm | |
| 25 mm | |
| Opaque | |
| 3 in. | |
| 3 x 1 in. (75 x 25 mm) |
| SuperFrost | |
| 76 mm | |
| White | |
| Superfrost Slide, 1.2mm thick | |
| 1 in. |