Learn More
Fisherbrand™ Sterile Microcentrifuge Tubes with Screw Caps
GREENER_CHOICE
Note: Image is representative only. It shows colored caps, but all of the tubes in this offering come with Natural caps; there are no color cap options.
 View more versions of this product
Catalog No. 02681372
Only null left
Description
Description
Choose skirted or conical-bottom style
  • With attached caps
  • Natural-colored screw caps with O-rings
  • RNase-free and DNase-free
Specifications
Chemical Identifiers
Polypropylene
Polypropylene
Sterile
Natural
O-ring
Screw Cap
Specifications
Polypropylene
Natural
O-ring
Screw Cap
18,000 x g
1.5 mL
Polypropylene
Sterile
Skirted
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories