Learn More
Fisherbrand™ Stainless-Steel Hydrolab Manifolds
For routine microbiological filtrations
Includes:
- Manifold with hose connector and pipe plug; manifold handles; three-way valve assemblies with Type 316 stainless-steel bodies
- Viton™ rubber O-rings and funnel holders.
Description
For routine microbiological filtrations
- Support microanalysis funnels when multiple samples are run from one vacuum line
- Made from polished Type 304 stainless steel
- Anodized aluminum handles on both ends for positioning on benchtop
- Sturdy units have low center of gravity to avoid tipping
- Each branch is operated by three-way valve to permit attachment of vent filter
- Accept vacuum funnel using a No. 8 stopper
- Autoclavable
- Serrated vacuum hose connection accepts 0.37 in. (9.5mm) tubing from an appropriate vacuum source
Specifications
| Autoclavable | |
| Hydrolab Manifold | |
| 6 in. | |
| 16.51 cm |
| Stainless Steel (304) | |
| 18 in. | |
| 6.5 in. |