missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stack and Nest Totes

Reusable container ideal for storing, work-in process and transporting items

$159.87

Specifications

Product Type Storage Bin (Stack and Nest Tote)
Color Blue
View More Specs

Products 1
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity & Availability  
15000210
View Documents Promotion Details
Fisherbrand™
FBSNT180BL
18 x 11 x 6 in.
Case of 6 Each for $159.87
Only null left
 
Description

Description

  • Available with or without lids
  • Stackable or can be turned 180 degrees to nest when empty, saving valuable space
  • Made of industrial-grade polypropylene
  • Do not rust, corrode or bend
Specifications

Specifications

Storage Bin (Stack and Nest Tote)
Blue
Videos
SDS
Documents

Documents