missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MicroMat™ CLR Silicone Mats for Well Plates

Safely seal plates and eliminate cross-contamination of samples with Fisherbrand™ MicroMat™ CLR Silicone Mats for Well Plates. Manufactured of pure silicone these mats resist coring and tearing and provide superior resealability after multiple injections.

$409.09 - $432.91

Specifications

Color Clear
Solvent Or Alcohol Resistant No Resistance
Material Silicone
Autoclavable Autoclavable
No. per Pack 10
View More Specs

Products 3
Catalog Number Mfr. No. Plate Compatibility Well Shape Pre-slit Includes Price Quantity & Availability  
Catalog Number Mfr. No. Plate Compatibility Well Shape Pre-slit Includes Price Quantity & Availability  
0339649
View Documents Promotion Details
Fisherbrand™
0339649
96-well Plate Round No Small Penetration Area
Pack of 10 Each for $409.09
Only null left
 
0339652
View Documents Promotion Details
Fisherbrand™
0339652
96-well Plate Round Yes Small Penetration Area
Pack of 10 Each for $432.91
Only null left
 
0339650
View Documents Promotion Details
Fisherbrand™
0339650
96-well Plate Round Yes Large Penetration Area
Pack of 10 Each for $424.94
Only null left
 
Description

Description

Safely seal plates and eliminate cross-contamination of samples with Fisherbrand™ MicroMat™ CLR Silicone Mats for Well Plates. Manufactured of pure silicone these mats resist coring and tearing and provide superior resealability after multiple injections.

Excellent Chemical and Thermal Compatibility

  • Solvent-resistant with excellent chemical compatibility
  • Dry-heat autoclavable and can withstand low temperatures to -80°C

Automation-Ready

  • Footprint allows plates to be securely stacked without robotic arm interference
  • Pre-slit mats offer better piercability and reduced vacuum formation
Specifications

Specifications

Clear
Silicone
10
Fisherbrand
No Resistance
Autoclavable
Sealing Mat
-80°C
Videos
SDS
Documents

Documents