missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Selectfrost™ Microscope Slides

Catalog No. 1255005
Change view
Click to view available options
Color:
Aqua
Blue
Burnt Orange
Green
Lilac
Pink
Tan
White
Yellow
Corner Style:
Clipped
Plain
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Corner Style
1255005 Aqua Plain
1255001 Blue Plain
1255006 Burnt Orange Plain
1255002 Green Plain
1255007 Lilac Plain
1255003 Pink Plain
1255008 Tan Plain
1255004 Yellow Plain
12550003 White Plain
22037205 White Clipped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 1255005

Color coded for easy identification

The Selectfrost microscope slides include colored tabs that allow grouping according to urgency, test, hospital or physician.

  • Autoclavable
  • Available in an array of tab colors
  • Durable, opaque tab is inert to common laboratory chemicals and reagents
  • Does NOT include SPECIMEN orientation key printed below tab
  • Packaged in paperboard box
  • Size: 25 × 75 × 1mm

Specifications

Length (Metric) 25 mm
Frosted Selectfrost Color-coded
Width (Metric) 75 mm
Thickness (Metric) 1 mm
Type Microscope Slide
Color Aqua
Corner Style Plain
Quantity 144/Pk.