Learn More
Fisher Healthcare™ Safetex™ No Touch™ Polypropylene Slide Holders
Patented window design allows for easy patient identification and helps ensure that slides reach the lab intact and unbroken
Supplier: Fisher Healthcare™ 880041
Description
- Safetex plastic
- Available in one- and two-slide configurations
- Sturdy polypropylene
- No Touch security prevents the specimen from touching the slide holder cover
- Holds one or two 3 x 1 in. slides
Specifications
| Polypropylene | |
| 2 Slides |