missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Microtube Support Racks

Excellent for microbiology work or any work using microtubes

$131.67

Specifications

Autoclavable Autoclavable
Color White
Material Polypropylene
Positions 72
Shape Rectangular
View More Specs

Products 1
Catalog Number Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Dimensions (L x W x H) Price Quantity & Availability  
032132
View Documents Promotion Details
8.37 x 4.37 x 4.37 in.
Each for $131.67
Case of 4 Each for $526.68
Only null left
 
Description

Description

  • Hold 72 micro sample tubes
  • Stackable
  • End grips for easy handling
  • Autoclavable to 121°C (250°F)
Specifications

Specifications

Autoclavable
Polypropylene
Rectangular
4/Cs.
Microtube Rack
White
72
Non-sterile
4.37 in.
Videos
SDS
Documents

Documents