Learn More
Fisherbrand™ Plastic Joint Clips
Made of plastic for use on standard taper glass joints
Supplier: Fisherbrand™ 14955258
Description
Made of plastic for use on standard taper glass joints
- Color coded for easy size identification
- Does not corrode
- Will not scratch glass
- Withstands temperatures up to 150°C
Chemical Identifiers
| 10/Pk. | |
| Joint Clip | |
| Plastic |
Specifications
| 10/Pk. | |
| Plastic | |
| Yellow |
| Joint Clip | |
| Joint Size 40 |