Learn More
Fisherbrand™ Plastic Joint Clips
Made of plastic for use on standard taper glass joints
Supplier:  Fisherbrand™ 14955254
 View more versions of this product
Catalog No. 14955254
Only null left
Description
Description
Made of plastic for use on standard taper glass joints
  • Color coded for easy size identification
  • Does not corrode
  • Will not scratch glass
  • Withstands temperatures up to 150°C
Specifications
Chemical Identifiers
10/Pk.
Joint Clip
Plastic
Specifications
10/Pk.
Plastic
Blue
Joint Clip
Joint Size 19
Product Suggestions
Product Suggestions