Learn More
Fisher Science Education™ Plant Cells Basic Slide Set
Take a closer look at plant cells.
Supplier:  Fisher Science Education™ S68787
Catalog No. S68787
Only null left
Includes:
Plastic box for 25 microscope slides
Description
Description
Set of six slides includes:
  • Mitosis, l.s. from Allium root tips showing all stages of plant mitosis carefully stained with iron-hematoxyline after Heidenhain
  • Epidermal cells of Allium cepa (onion), flat mount shows typical plant cells with nuclei, cytoplasm and cell walls
  • Sieve plates in top view, t.s. of Cucurbita stem showing large structures
  • Spirogyra, in scalariform conjugation and after the stage of conjugation showing zygotes, w.m.
  • Wood cells, fibers and vessels of Fagus, beech, macerated and w.m.
  • Starch grains, sec. of tuber of Solanum tuberosum, potato showing starch and cork cells
Specifications
Specifications
Plant Cells Basic Slide Set
Product Suggestions
Product Suggestions